missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-pyrrolidinone, 97%, Thermo Scientific Chemicals

Product Code. 11471680
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101385-93-7
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD01631194
InChI Key JSOMVCDXPUXKIC-UHFFFAOYSA-N
Synonym n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
PubChem CID 471360
IUPAC Name tert-butyl 3-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C1

Specifications

Melting Point 34°C to 38°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material 1-Boc-3-pyrrolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'