missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-piperidinemethanol, 97%, Thermo Scientific Chemicals

36.10 € - 117.00 €

Chemical Identifiers

CAS 123855-51-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD02094488
InChI Key CTEDVGRUGMPBHE-UHFFFAOYSA-N
Synonym n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol
PubChem CID 2764081
IUPAC Name tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CO)CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362549
View Documents
Thermo Scientific Alfa Aesar
H26650.03
1 g
36.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
11372549
View Documents
Thermo Scientific Alfa Aesar
H26650.06
5 g
117.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123855-51-6
215.29
CTEDVGRUGMPBHE-UHFFFAOYSA-N
2764081
CC(C)(C)OC(=O)N1CCC(CO)CC1
C11H21NO3
MFCD02094488
n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol
tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate

Specifications

123855-51-6
C11H21NO3
n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol
CC(C)(C)OC(=O)N1CCC(CO)CC1
215.29
215.29
1-Boc-4-piperidinemethanol
78°C to 82°C
MFCD02094488
CTEDVGRUGMPBHE-UHFFFAOYSA-N
tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
2764081
97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.