Learn More
1-(Boc-amino)cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals
26.00 € - 78.70 €
Chemical Identifiers
CAS | 88950-64-5 |
---|---|
Molecular Formula | C9H15NO4 |
Molecular Weight (g/mol) | 201.22 |
MDL Number | MFCD00083257 |
InChI Key | DSKCOVBHIFAJRI-UHFFFAOYSA-N |
Synonym | 1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino |
PubChem CID | 688396 |
IUPAC Name | 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid |
SMILES | CC(C)(C)OC(=O)NC1(CC1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458588
|
Thermo Scientific Alfa Aesar
H61992.MD |
250 mg | N/A | ||||||
15448588
|
Thermo Scientific Alfa Aesar
H61992.03 |
1 g |
78.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88950-64-5 | |
201.22 | |
DSKCOVBHIFAJRI-UHFFFAOYSA-N | |
688396 | |
CC(C)(C)OC(=O)NC1(CC1)C(O)=O |
C9H15NO4 | |
MFCD00083257 | |
1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid |
Specifications
118°C (decomposition) | |
C9H15NO4 | |
4430628 | |
DSKCOVBHIFAJRI-UHFFFAOYSA-N | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopropane-1-carboxylic acid | |
688396 | |
98% |
88950-64-5 | |
MFCD00083257 | |
1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino | |
CC(C)(C)OC(=O)NC1(CC1)C(O)=O | |
201.22 | |
201.22 | |
1-(Boc-amino)cyclopropanecarboxylic acid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.