missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-N-Fmoc-D-tryptophan, 98%, Thermo Scientific Chemicals

Product Code. 15478738
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163619-04-3
Molecular Formula C31H30N2O6
Molecular Weight (g/mol) 526.59
MDL Number MFCD00153367
InChI Key ADOHASQZJSJZBT-AREMUKBSSA-N
Synonym fmoc-d-trp boc-oh, n, a-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc, ambotzfaa1339, pubchem10051, 1-boc-fmoc-d-tryptophan, 1-boc-n-fmoc-d-tryptophan, 2r-3-1-tert-butoxycarbonylindol-3-yl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, nalpha-fmoc-n in-boc-d-tryptophan, fmoc-d-trp boc-oh hplc
PubChem CID 16213150
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]indol-3-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12

Specifications

Quantity 250 mg
Beilstein 7062970
Formula Weight 526.59
Percent Purity 98%
Chemical Name or Material 1-Boc-N-Fmoc-D-tryptophan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'