Learn More
1-Boc-piperazine, 99%, Thermo Scientific Chemicals
Description
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.
Solubility
Soluble in ethyl acetate, methanol and water.
Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 44°C to 48°C |
Quantity | 25 g |
Beilstein | 879985 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in ethyl acetate,methanol and water. |
Formula Weight | 186.26 |
Percent Purity | 99% |
Chemical Name or Material | 1-Boc-piperazine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X11C002 | Certificate of Analysis | 25/11/2024 | L13363.06,L13363.03,L13363.14 |
10242524 | Certificate of Analysis | 04/08/2022 | L13363.06,L13363.03,L13363.14 |
X11C002 | Certificate of Analysis | 08/12/2021 | L13363.06,L13363.03,L13363.14 |
10202451 | Certificate of Analysis | 07/12/2021 | L13363.06,L13363.03,L13363.14 |
61001114 | Certificate of Analysis | 07/12/2021 | L13363.06,L13363.03,L13363.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.