missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dimethoxybenzene, 98%, Thermo Scientific Chemicals

67.40 € - 910.00 €

Chemical Identifiers

CAS 17715-69-4
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00009844
InChI Key NIUZVSQOXJIHBL-UHFFFAOYSA-N
Synonym 2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
PubChem CID 87266
IUPAC Name 1-bromo-2,4-dimethoxybenzene
SMILES COC1=CC=C(Br)C(OC)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11427087
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14439.14
25 g
67.40 €
25g
Please to purchase this item. Need a web account? Register with us today!
11437087
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14439.22
100 g
206.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
11447087
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14439.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17715-69-4
217.06
NIUZVSQOXJIHBL-UHFFFAOYSA-N
87266
COC1=CC=C(Br)C(OC)=C1
C8H9BrO2
MFCD00009844
2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
1-bromo-2,4-dimethoxybenzene

Specifications

17715-69-4
1.496
113°C (235°F)
1.572
1867593
NIUZVSQOXJIHBL-UHFFFAOYSA-N
1-bromo-2,4-dimethoxybenzene
87266
98%
24°C to 26°C
153°C to 155°C (18 mmHg)
C8H9BrO2
MFCD00009844
2,4-dimethoxybromobenzene, 1,3-dimethoxy-4-bromobenzene, 4-bromo-3-methoxyanisole, benzene, 1-bromo-2,4-dimethoxy, 4-bromoresorcinol dimethyl ether, 4-bromoresocinol dimethyl ether, 4-bromo-1,3-dimethoxybenzene, 1-brom-2,4-dimethoxybenzol, pubchem3036, acmc-1c5dw
COC1=CC=C(Br)C(OC)=C1
217.06
217.07
1-Bromo-2,4-dimethoxybenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 241-717-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only