missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Product Code. 11439360
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37763-43-2
Molecular Formula C11H8Br2
Molecular Weight (g/mol) 299.99
MDL Number MFCD00046369
InChI Key DQTOCXIHYIQHCK-UHFFFAOYSA-N
Synonym 1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene
PubChem CID 37828
IUPAC Name 1-bromo-2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 108°C
Quantity 250 mg
UN Number UN3261
Formula Weight 299.99
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-(bromomethyl)naphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'