Learn More
1-Bromo-2-(bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
67.00 € - 239.00 €
Chemical Identifiers
CAS | 37763-43-2 |
---|---|
Molecular Formula | C11H8Br2 |
Molecular Weight (g/mol) | 299.99 |
MDL Number | MFCD00046369 |
InChI Key | DQTOCXIHYIQHCK-UHFFFAOYSA-N |
Synonym | 1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene |
PubChem CID | 37828 |
IUPAC Name | 1-bromo-2-(bromomethyl)naphthalene |
SMILES | BrCC1=CC=C2C=CC=CC2=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439360
|
Thermo Scientific Alfa Aesar
H50240.MD |
250 mg | |||||||
11449360
|
Thermo Scientific Alfa Aesar
H50240.03 |
1 g |
239.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37763-43-2 | |
299.99 | |
DQTOCXIHYIQHCK-UHFFFAOYSA-N | |
37828 | |
BrCC1=CC=C2C=CC=CC2=C1Br |
C11H8Br2 | |
MFCD00046369 | |
1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene | |
1-bromo-2-(bromomethyl)naphthalene |
Specifications
37763-43-2 | |
C11H8Br2 | |
UN3261 | |
DQTOCXIHYIQHCK-UHFFFAOYSA-N | |
1-bromo-2-(bromomethyl)naphthalene | |
37828 | |
98% |
108°C | |
MFCD00046369 | |
1-bromo-2-bromomethyl naphthalene, naphthalene, 1-bromo-2-bromomethyl, 1-bromo-2-bromomethyl-naphthalene, acmc-209ivc, bromobromomethylnaphtalene, 3-05-00-01633 beilstein handbook reference, wln: l66j be c1e, 1-bromo-2-bromethylnaphthalene, 2-bromomethyl-1-bromonaphthalene | |
BrCC1=CC=C2C=CC=CC2=C1Br | |
299.99 | |
299.99 | |
1-Bromo-2-(bromomethyl)naphthalene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : QJ1550000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only