Learn More
1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals
41.10 € - 523.61 €
Chemical Identifiers
CAS | 107-04-0 |
---|---|
Molecular Formula | C2H4BrCl |
Molecular Weight (g/mol) | 143.408 |
MDL Number | MFCD00000962 |
InChI Key | IBYHHJPAARCAIE-UHFFFAOYSA-N |
Synonym | ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane |
PubChem CID | 7849 |
ChEBI | CHEBI:19032 |
IUPAC Name | 1-bromo-2-chloroethane |
SMILES | C(CBr)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11469473
|
Thermo Scientific Alfa Aesar
A12349.22 |
100 g |
41.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479473
|
Thermo Scientific Alfa Aesar
A12349.36 |
500 g |
143.72 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489473
|
Thermo Scientific Alfa Aesar
A12349.0E |
2500 g |
523.61 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.
Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.
Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
107-04-0 | |
143.408 | |
IBYHHJPAARCAIE-UHFFFAOYSA-N | |
7849 | |
1-bromo-2-chloroethane |
C2H4BrCl | |
MFCD00000962 | |
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
CHEBI:19032 | |
C(CBr)Cl |
Specifications
107-04-0 | |
1.726 | |
None | |
C2H4BrCl | |
1.49 | |
UN2810 | |
Light sensitive | |
Soluble in water ( 7g/L (20°C). Soluble in ethanol,ethyl ether,chloroform. | |
C(CBr)Cl | |
143.408 | |
CHEBI:19032 | |
98% |
-18°C | |
106°C to 107°C | |
Sweet | |
ClCH2CH2Br | |
MFCD00000962 | |
605265 | |
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
IBYHHJPAARCAIE-UHFFFAOYSA-N | |
1-bromo-2-chloroethane | |
7849 | |
143.42 | |
1-Bromo-2-chloroethane |
Sicherheit und Handhabung
GHS H Statement
H301-H350-H312-H332-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335-H341
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 203-456-0
missing translation for 'rtecsNumber' : KH6500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only