missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals Product Code.: 11433364

1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11433364
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11433364

Brand: Thermo Scientific Alfa Aesar A10635.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br

Specifications

Melting Point -8°C
Density 1.601
Boiling Point 155°C to 157°C
Flash Point 43°C (109°F)
Odor Aromatic
Refractive Index 1.534
Quantity 500 g
UN Number UN1993
Beilstein 1071819
Solubility Information Insoluble in water.
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-2-fluorobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237449 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10635.22,A10635.14,A10635.36
Lot Number5005F02W Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10635.22,A10635.14,A10635.36
Lot Number10138140 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10635.22,A10635.14,A10635.36
Lot Number10198737 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10635.22,A10635.14,A10635.36
Lot Number10220497 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10635.22,A10635.14,A10635.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-2-fluorobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.