missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methylnaphthalene, tech. 90%, Thermo Scientific Chemicals

Product Code. 11331827
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methylnaphthalene is used as test compound in determination of halogenated hydrocarbons at trace levels by supercritical fluid chromatography-microwave-induced plasma mass spectrometry. Dynamic phosphorescence quenching of 1-bromo-2-methylnaphthalene without deoxygenation has been used for selective sensing of Cu(II) at ngm levels.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2586-62-1
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00003871
InChI Key CMIMBQIBIZZZHQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo-2-methyl, 2-methyl-1-bromonaphthalene, .beta.-methyl-.alpha.-bromonaphthalene, zlchem 421, 1-bromo-2-methylnaphtalene, acmc-1cq62, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromonaphthalen-2-yl methyl
PubChem CID 75754
IUPAC Name 1-bromo-2-methylnaphthalene
SMILES CC1=CC=C2C=CC=CC2=C1Br

Specifications

Density 1.428
Boiling Point 295°C to 296°C
Flash Point >110°C (230°F)
Refractive Index 1.649
Quantity 10 g
Beilstein 2042531
Solubility Information Not miscible in water.
Formula Weight 221.1
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Bromo-2-methylnaphthalene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'