missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10096293
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-19-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007045
InChI Key ORPVVAKYSXQCJI-UHFFFAOYSA-N
Synonym 2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro
PubChem CID 11341
IUPAC Name 1-bromo-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 40.0°C to 43.0°C
Boiling Point 261.0°C
Flash Point >110°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NO2
Quantity 5 g
Beilstein 05, I, 247
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-nitrobenzene, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.