Learn More
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals
22.80 € - 236.00 €
Chemical Identifiers
CAS | 109-70-6 |
---|---|
Molecular Formula | C3H6BrCl |
Molecular Weight (g/mol) | 157.435 |
MDL Number | MFCD00000998 |
InChI Key | MFESCIUQSIBMSM-UHFFFAOYSA-N |
Synonym | trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane |
PubChem CID | 8006 |
IUPAC Name | 1-bromo-3-chloropropane |
SMILES | C(CCl)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484190
|
Thermo Scientific Alfa Aesar
A11395.22 |
100 g |
22.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440223
|
Thermo Scientific Alfa Aesar
A11395.30 |
250 g |
27.60 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494190
|
Thermo Scientific Alfa Aesar
A11395.36 |
500 g |
41.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430223
|
Thermo Scientific Alfa Aesar
A11395.0B |
1000 g |
62.30 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450223
|
Thermo Scientific Alfa Aesar
A11395.0I |
5000 g |
236.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.
Solubility
Miscible with alcohols, chlorofom and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.
Chemical Identifiers
109-70-6 | |
157.435 | |
MFESCIUQSIBMSM-UHFFFAOYSA-N | |
8006 | |
C(CCl)CBr |
C3H6BrCl | |
MFCD00000998 | |
trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane | |
1-bromo-3-chloropropane |
Specifications
109-70-6 | |
1.598 | |
>140°C (284°F) | |
1.4855 | |
UN2688 | |
trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane | |
MFESCIUQSIBMSM-UHFFFAOYSA-N | |
1-bromo-3-chloropropane | |
8006 | |
99% |
-59°C | |
142°C to 145°C | |
C3H6BrCl | |
MFCD00000998 | |
605278 | |
Miscible with alcohols,chlorofom and ether. Immiscible with water. | |
C(CCl)CBr | |
157.435 | |
157.44 | |
1-Bromo-3-chloropropane |
Safety and Handling
GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P311-P330-P370+P378q-P501c
H227-H302-H331-H335-H341
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1-BROMO-3-CHLOROPROPANE
missing translation for 'einecsNumber' : 203-697-1
missing translation for 'rtecsNumber' : TX4113000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only