missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane, 98%, Thermo Scientific Chemicals

Product Code. 11454190
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-methylbutane was used in the synthesis of 1-(3-methylbutyl)pyrrole and in the production of pentyl peroxy radical by direct photolysis production method. It was also used as extraction solvent for determination of polycyclic aromatic hydrocarbons in water samples using liquid-liquid microextraction combined with gas chromatography-mass spectrometry. It is also used as an alkyl halide and organic synthesis reagent.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000253
InChI Key YXZFFTJAHVMMLF-UHFFFAOYSA-N
Synonym isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
PubChem CID 7891
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(C)CCBr

Specifications

Melting Point -112°C
Density 1.208
Boiling Point 120°C to 121°C
Flash Point 22°C (71°F)
Refractive Index 1.4409
Quantity 5000 g
UN Number UN2341
Beilstein 773652
Merck Index 14,5114
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 151.05
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-methylbutane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.