missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenoxypropane, 98%, Thermo Scientific Chemicals

Product Code. 11333186
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-63-6
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00000256
InChI Key NIDWUZTTXGJFNN-UHFFFAOYSA-N
Synonym 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide
PubChem CID 68522
IUPAC Name 3-bromopropoxybenzene
SMILES BrCCCOC1=CC=CC=C1

Specifications

Melting Point 10°C to 11°C
Density 1.365
Boiling Point 130°C to 134°C (14 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.546
Quantity 25 g
Beilstein 508978
Formula Weight 215.1
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenoxypropane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'