missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals

Product Code. 11477956
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

Solubility
Soluble in water (partly). Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name 3-bromopropylbenzene
SMILES BrCCCC1=CC=CC=C1

Specifications

Density 1.308
Boiling Point 219°C to 220°C
Flash Point 101°C (213°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 2205527
Solubility Information Soluble in water (partly). Soluble in chloroform and methanol.
Formula Weight 199.09
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenylpropane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'