missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-butylbenzene, 97%, Thermo Scientific Chemicals

43.80 € - 472.00 €

Chemical Identifiers

CAS 41492-05-1
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00040934
InChI Key BRGVKVZXDWGJBX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
PubChem CID 521059
IUPAC Name 1-bromo-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)Br
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10682752
View Documents Promotion Details
Thermo Scientific
320440100
10 g Glass bottle
43.80 €
10g
Please to purchase this item. Need a web account? Register with us today!
10400283
View Documents Promotion Details
Thermo Scientific Acros
320440500
50 g Glass bottle
130.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
10239413
View Documents Promotion Details
Thermo Scientific Acros
320442500
250 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

41492-05-1
213.12
BRGVKVZXDWGJBX-UHFFFAOYSA-N
521059
CCCCC1=CC=C(C=C1)Br
C10H13Br
MFCD00040934
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
1-bromo-4-butylbenzene

Specifications

41492-05-1
100.0
1.2000g/mL
Authentic
C10H13Br
1.5300 to 1.533
1.2
BRGVKVZXDWGJBX-UHFFFAOYSA-N
1-bromo-4-butylbenzene
521059
97%
1-Bromo-4-butylbenzene
96.0
Colorless to Yellow
108°C
96% min. (GC)
BrC6H4(CH2)3CH3
MFCD00040934
1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
CCCCC1=CC=C(C=C1)Br
213.12
213.12
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only