missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%

Product Code. p-72621 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10607252

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6940-78-9 | C4H8BrCl | 171.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.46
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.4800g/mL
Boiling Point 80.0°C to 82.0°C (30.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)4Br
Refractive Index 1.4865 to 1.4885
Quantity 25 g
Specific Gravity 1.48
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with organic solvents,soluble in ethanol,ethylether,slightly soluble in carbontetrachloride
Formula Weight 171.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-chlorobutane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-4-chlorobutane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.