missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(difluoromethoxy)benzene, 97%, Thermo Scientific Chemicals

Product Code. 11395289
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5905-69-1
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00221465
InChI Key ORIYZUFTROJBQJ-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethoxy benzene, 4-difluoromethoxy bromobenzene, p-difluoromethoxybromobenzene, 1-difluoromethoxy-4-bromobenzene, benzene, 1-bromo-4-difluoromethoxy, l-bromo-4-difluoromethoxy benzene, pubchem4309, acmc-209m9x, p-difluoro-methoxybromobenzene, ksc495s1h
PubChem CID 2737007
IUPAC Name 1-bromo-4-(difluoromethoxy)benzene
SMILES C1=CC(=CC=C1OC(F)F)Br

Specifications

Density 1.631
Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.501
Quantity 5 g
UN Number UN1993
Beilstein 2521410
Formula Weight 223.02
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(difluoromethoxy)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'