missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoronaphthalene, 98%, Thermo Scientific Chemicals

38.90 € - 403.00 €

Chemical Identifiers

CAS 341-41-3
Molecular Formula C10H6BrF
Molecular Weight (g/mol) 225.06
MDL Number MFCD00051473
InChI Key VAUJZKBFENPOCH-UHFFFAOYSA-N
Synonym 4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
PubChem CID 67647
IUPAC Name 1-bromo-4-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444190
View Documents
Thermo Scientific Alfa Aesar
A16934.03
1 g
38.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
11448766
View Documents
Thermo Scientific Alfa Aesar
A16934.06
5 g
125.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11458766
View Documents
Thermo Scientific Alfa Aesar
A16934.14
25 g
403.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

341-41-3
225.06
VAUJZKBFENPOCH-UHFFFAOYSA-N
67647
C1=CC=C2C(=C1)C(=CC=C2Br)F
C10H6BrF
MFCD00051473
4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
1-bromo-4-fluoronaphthalene

Specifications

341-41-3
>110°C (230°F)
MFCD00051473
4-bromo-1-fluoronaphthalene, 1-fluoro-4-bromonaphthalene, naphthalene, 1-bromo-4-fluoro, acmc-1cnye, 1-bromo 4-fluoronaphthalene, 1-bromo-4-fluoro naphthalene, 1-bromo-4-fluoro-naphthalene, ksc493c4t, 1-bromo-4-fluoronaphthalene
C1=CC=C2C(=C1)C(=CC=C2Br)F
225.06
225.06
1-Bromo-4-fluoronaphthalene
33°C to 37°C
C10H6BrF
2085804
VAUJZKBFENPOCH-UHFFFAOYSA-N
1-bromo-4-fluoronaphthalene
67647
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 206-434-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only