Learn More
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
34.27 € - 148.74 €
Chemical Identifiers
CAS | 586-78-7 |
---|---|
Molecular Formula | C6H4BrNO2 |
Molecular Weight (g/mol) | 202.01 |
MDL Number | MFCD00007280 |
InChI Key | ZDFBKZUDCQQKAC-UHFFFAOYSA-N |
Synonym | 4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115Show More |
PubChem CID | 11466 |
IUPAC Name | 1-bromo-4-nitrobenzene |
SMILES | [O-][N+](=O)C1=CC=C(Br)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10664662
|
Thermo Scientific Acros
173360100 |
10 g | Glass bottle |
34.27 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499300
|
Thermo Scientific Acros
173360250 |
25 g | Plastic bottle |
57.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409310
|
Thermo Scientific Acros
173361000 |
100 g | Plastic bottle |
148.74 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
586-78-7 | |
202.01 | |
ZDFBKZUDCQQKAC-UHFFFAOYSA-N | |
11466 | |
[O-][N+](=O)C1=CC=C(Br)C=C1 |
C6H4BrNO2 | |
MFCD00007280 | |
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115Show More | |
1-bromo-4-nitrobenzene |
Specifications
586-78-7 | |
100.0 | |
Yellow | |
Authentic | |
C6H4BrNO2 | |
MFCD00007280 | |
4-bromonitrobenzene, 4-nitrobromobenzene, p-bromonitrobenzene, benzene, 1-bromo-4-nitro, p-nitrobromobenzene, 4-nitrophenyl bromide, 4-bromo-1-nitrobenzene, p-nitrophenyl bromide, 1-bromo-4-nitro-benzene, ccris 3115 | |
ZDFBKZUDCQQKAC-UHFFFAOYSA-N | |
1-bromo-4-nitrobenzene | |
11466 | |
98% | |
1-Bromo-4-nitrobenzene, 99% |
98.5 | |
125.0°C to 127.0°C | |
255.0°C to 256.0°C | |
97.5% min. (GC) | |
BrC6H4NO2 | |
05, 248 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether | |
[O-][N+](=O)C1=CC=C(Br)C=C1 | |
202.01 | |
202.01 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-583-8
missing translation for 'rtecsNumber' : CY9040550
missing translation for 'tsca' : TSCA
- 1-Bromo-4-nitrobenzene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.