missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-phenylbutane, 98%, Thermo Scientific Chemicals
28.50 € - 125.00 €
Chemical Identifiers
CAS | 13633-25-5 |
---|---|
Molecular Formula | C10H13Br |
Molecular Weight (g/mol) | 213.12 |
MDL Number | MFCD00154988 |
InChI Key | XPBQQAHIVODAIC-UHFFFAOYSA-N |
Synonym | 1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane |
PubChem CID | 259668 |
SMILES | BrCCCCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490758
|
Thermo Scientific Alfa Aesar
B25237.03 |
1 g |
28.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400768
|
Thermo Scientific Alfa Aesar
B25237.06 |
5 g |
125.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemische Identifikatoren
13633-25-5 | |
213.12 | |
XPBQQAHIVODAIC-UHFFFAOYSA-N | |
259668 |
C10H13Br | |
MFCD00154988 | |
1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
BrCCCCC1=CC=CC=C1 |
Spezifikation
13633-25-5 | |
131°C to 133°C (12 mmHg) | |
MFCD00154988 | |
Slightly soluble in water. | |
BrCCCCC1=CC=CC=C1 | |
213.12 | |
213.12 | |
1-Bromo-4-phenylbutane |
1.214 | |
C10H13Br | |
1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
XPBQQAHIVODAIC-UHFFFAOYSA-N | |
(4-bromobutyl)benzene | |
259668 | |
98% |
Sicherheit und Handhabung
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only