missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals

Product Code. 10003913
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4000g/mL
Boiling Point 210°C to 212°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Cl(CH2)5Br
Refractive Index 1.4826 to 1.4846
Quantity 100 g
Beilstein 01,III,346
Specific Gravity 1.4
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in methanol,ethanol,chloroform,and ether
Formula Weight 185.49
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-5-chloropentane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-5-chloropentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'