missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-6-phenylhexane, 97%, Thermo Scientific Chemicals

43.60 € - 473.00 €

Chemical Identifiers

CAS 27976-27-8
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD00236022
InChI Key RAOLIGFNQJMMKW-UHFFFAOYSA-N
Synonym 6-bromohexyl benzene, 1-bromo-6-phenylhexane, benzene, 6-bromohexyl, 1-bromo-6-phenyl-hexane, 6-phenylhexyl bromide, hexane, 1-bromo-6-phenyl, 6-bromanylhexylbenzene, 5-benzylpentyl bromide, 6-bromo-1-phenylhexane
PubChem CID 561788
IUPAC Name 6-bromohexylbenzene
SMILES C1=CC=C(C=C1)CCCCCCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333596
View Documents
Thermo Scientific Alfa Aesar
L14568.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313596
View Documents
Thermo Scientific Alfa Aesar
L14568.03
1 g
124.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11323596
View Documents
Thermo Scientific Alfa Aesar
L14568.06
5 g
473.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27976-27-8
241.172
RAOLIGFNQJMMKW-UHFFFAOYSA-N
561788
C1=CC=C(C=C1)CCCCCCBr
C12H17Br
MFCD00236022
6-bromohexyl benzene, 1-bromo-6-phenylhexane, benzene, 6-bromohexyl, 1-bromo-6-phenyl-hexane, 6-phenylhexyl bromide, hexane, 1-bromo-6-phenyl, 6-bromanylhexylbenzene, 5-benzylpentyl bromide, 6-bromo-1-phenylhexane
6-bromohexylbenzene

Specifications

27976-27-8
C12H17Br
MFCD00236022
6-bromohexyl benzene, 1-bromo-6-phenylhexane, benzene, 6-bromohexyl, 1-bromo-6-phenyl-hexane, 6-phenylhexyl bromide, hexane, 1-bromo-6-phenyl, 6-bromanylhexylbenzene, 5-benzylpentyl bromide, 6-bromo-1-phenylhexane
C1=CC=C(C=C1)CCCCCCBr
241.172
241.18
1-Bromo-6-phenylhexane
115°C to 118°C (6 mmHg)
1.5295
2207649
RAOLIGFNQJMMKW-UHFFFAOYSA-N
6-bromohexylbenzene
561788
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.