missing translation for 'onlineSavingsMsg'
Learn More

1-Bromodecane, 98%, Thermo Scientific Chemicals

26.83 € - 730.64 €

Chemical Identifiers

CAS 112-29-8
Molecular Formula C10H21Br
Molecular Weight (g/mol) 221.18
MDL Number MFCD00000221
InChI Key MYMSJFSOOQERIO-UHFFFAOYSA-N
Synonym decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
PubChem CID 8173
IUPAC Name 1-bromodecane
SMILES CCCCCCCCCCBr
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11414190
View Documents
Thermo Scientific Alfa Aesar
A11340.22
100 g
26.83 €
100g
Please to purchase this item. Need a web account? Register with us today!
11433754
View Documents
Thermo Scientific Alfa Aesar
A11340.36
500 g
58.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
11443754
View Documents
Thermo Scientific Alfa Aesar
A11340.0E
2500 g
214.07 €
2500g
Please to purchase this item. Need a web account? Register with us today!
11424190
View Documents
Thermo Scientific Alfa Aesar
A11340.0C
10,000 g
730.64 €
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromodecane is used to produce decanal. It is also used in organic synthesis.

Solubility
Miscible with most common organic solvents like ethanol, ethyl ether and chloroform. Not miscible in water.

Notes
Do not store together with strongly basic or oxidizing materials. Incompatible with strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

112-29-8
221.18
MYMSJFSOOQERIO-UHFFFAOYSA-N
8173
CCCCCCCCCCBr
C10H21Br
MFCD00000221
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
1-bromodecane

Specifications

112-29-8
1.067
94°C (201°F)
CH3(CH2)9Br
MFCD00000221
1735227
Miscible with most common organic solvents like ethanol,ethyl ether and chloroform. Not miscible in water.
CCCCCCCCCCBr
221.18
221.19
1-Bromodecane
-30°C
238°C
C10H21Br
1.456
UN2810
decyl bromide, n-decyl bromide, decane, 1-bromo, 1-decyl bromide, decylbromide, bromodecane, decane, bromo, labotest-bb ltbb001164, decylmonobromide, n-decylbromide
MYMSJFSOOQERIO-UHFFFAOYSA-N
1-bromodecane
8173
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331
Toxic if inhaled.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-955-3

missing translation for 'rtecsNumber' : HD6850000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Bromodecane

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 1-Bromodecane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.