missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododecane, 98%, Thermo Scientific Chemicals

Product Code. 10764211
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.23
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododecane
SMILES CCCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -11°C to 9°C
Density 1.0300g/mL
Boiling Point 134.0°C to 135.0°C (6.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11Br
Refractive Index 1.4570 to 1.4590
Quantity 1 L
Beilstein 01,II,133
Merck Index 15,5444
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble
Formula Weight 249.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromododecane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.