missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoheptane, 98+%
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
Description
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Immiscible with water.
Notes
Flammable and incompatible with oxidizing agents, strong bases.
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Immiscible with water.
Notes
Flammable and incompatible with oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | -58°C |
| Density | 1.139 |
| Boiling Point | 179°C to 180°C |
| Flash Point | 60°C (140°F) |
| Odor | Recognizable |
| Refractive Index | 1.4499 |
| Quantity | 1000 g |
| UN Number | UN1993 |
| Beilstein | 1697160 |
| Solubility Information | Immiscible with water. |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction