Learn More
1-Bromohexadecane, 98%, Thermo Scientific Chemicals
24.22 € - 135.68 €
Chemical Identifiers
CAS | 112-82-3 |
---|---|
Molecular Formula | C16H33Br |
Molecular Weight (g/mol) | 305.34 |
MDL Number | MFCD00000230 |
InChI Key | HNTGIJLWHDPAFN-UHFFFAOYSA-N |
Synonym | hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam |
PubChem CID | 8213 |
IUPAC Name | 1-bromohexadecane |
SMILES | CCCCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494180
|
Thermo Scientific Alfa Aesar
A15147.14 |
25 g |
24.22 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422128
|
Thermo Scientific Alfa Aesar
A15147.30 |
250 g |
48.84 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11412128
|
Thermo Scientific Alfa Aesar
A15147.0B |
1000 g |
135.68 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
112-82-3 | |
305.34 | |
HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
8213 | |
CCCCCCCCCCCCCCCCBr |
C16H33Br | |
MFCD00000230 | |
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
1-bromohexadecane |
Specifications
112-82-3 | |
1 | |
176°C (348°F) | |
C16H33Br | |
MFCD00000230 | |
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
1-bromohexadecane | |
8213 | |
98% |
16°C to 18°C | |
336°C | |
Odorless | |
1.462 | |
773989 | |
Difficult to mix. | |
CCCCCCCCCCCCCCCCBr | |
305.34 | |
305.35 | |
1-Bromohexadecane |
Safety and Handling
missing translation for 'einecsNumber' : 204-008-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.