missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)adamantane, 97%, Thermo Scientific™

37.97 € - 168.00 €

Chemical Identifiers

CAS 14651-42-4
Molecular Formula C11H17Br
Molecular Weight (g/mol) 229.161
InChI Key RLRBYCAVZBQANU-UHFFFAOYSA-N
Synonym 1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1∼3,7∼ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
PubChem CID 1416304
IUPAC Name 1-(bromomethyl)adamantane
SMILES C1C2CC3CC1CC(C2)(C3)CBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10509852
View Documents
Thermo Scientific Maybridge
BTB13836DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10396494
View Documents
Thermo Scientific Maybridge
BTB13836DE
5 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

14651-42-4
229.161
1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1∼3,7∼ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
1-(bromomethyl)adamantane
C11H17Br
RLRBYCAVZBQANU-UHFFFAOYSA-N
1416304
C1C2CC3CC1CC(C2)(C3)CBr

Specifications

14651-42-4
C11H17Br
RLRBYCAVZBQANU-UHFFFAOYSA-N
1-(bromomethyl)adamantane
1416304
97%
97%
1-bromomethyl adamantane, tricyclo 3.3.1.13,7 decane, 1-bromomethyl, adamantanylbromomethane, 1-bromomethyl tricyclo 3.3.1.1∼3,7∼ decane, 3r,5s,7s-1-bromomethyl adamantane, pubchem21524, 1-adamantylmethylbromide, 1-bromoethyl adamantane, 1-adamantyl methyl bromide, 1s,3r,5s,7s-1-bromomethyl adamantane
C1C2CC3CC1CC(C2)(C3)CBr
229.161
229.16
1-(Bromomethyl)adamantane
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.