Learn More
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
109.55 € - 409.00 €
Chemical Identifiers
CAS | 3163-27-7 |
---|---|
Molecular Formula | C11H9Br |
Molecular Weight (g/mol) | 221.097 |
MDL Number | MFCD00010804 |
InChI Key | RZJGKPNCYQZFGR-UHFFFAOYSA-N |
Synonym | 1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 |
PubChem CID | 137844 |
IUPAC Name | 1-(bromomethyl)naphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15471169
|
Thermo Scientific Alfa Aesar
H64116.14 |
25 g |
109.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15481169
|
Thermo Scientific Alfa Aesar
H64116.22 |
100 g | N/A | ||||||
Description
It is employed as a intermediate for organic synthesis and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
3163-27-7 | |
221.097 | |
RZJGKPNCYQZFGR-UHFFFAOYSA-N | |
137844 | |
C1=CC=C2C(=C1)C=CC=C2CBr |
C11H9Br | |
MFCD00010804 | |
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 | |
1-(bromomethyl)naphthalene |
Specifications
3163-27-7 | |
1.44 g/mL | |
C11H9Br | |
MFCD00010804 | |
742417 | |
Slightly soluble in water. | |
C1=CC=C2C(=C1)C=CC=C2CBr | |
221.097 | |
221.1 | |
Powder |
White | |
213°C (100 mmHg) | |
1.66 | |
UN3261 | |
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 | |
RZJGKPNCYQZFGR-UHFFFAOYSA-N | |
1-(bromomethyl)naphthalene | |
137844 | |
98% | |
1-(Bromomethyl)naphthalene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 801-817-2
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.