missing translation for 'onlineSavingsMsg'
Learn More

1-Bromonaphthalene, 96%

Product Code. 10488050 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10488050

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-11-9 | C10H7Br | 207.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-11-9, 580-13-2
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -1.0°C
Color Brown-Yellow to Yellow
Density 1.4890g/mL
Boiling Point 279.0°C to 281.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 2.5% max. 2-Bromonaphthalene (GC)
Packaging Glass bottle
Molecular Formula C10H7Br
Linear Formula C10H7Br
Refractive Index 1.6560 to 1.6590
Quantity 500 mL
Beilstein 05, 547
Merck Index 15, 1436
Specific Gravity 1.489
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether and ethanol
Formula Weight 207.07
Vapor Pressure 14.8hPa at 20°C
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromonaphthalene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Bromonaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.