missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane, 96%, Thermo Scientific Chemicals

15.50 € - 437.18 €

Chemical Identifiers

CAS 112-89-0
Molecular Formula C18H37Br
Molecular Weight (g/mol) 333.40
MDL Number MFCD00000231
InChI Key WSULSMOGMLRGKU-UHFFFAOYSA-N
Synonym octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
PubChem CID 8218
IUPAC Name 1-bromooctadecane
SMILES CCCCCCCCCCCCCCCCCCBr
View More Specs

Productos 4
Código de producto Marca Quantity Packaging Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Packaging Precio Cantidad y disponibilidad  
10467750
missing translation for 'viewDocuments'
Thermo Scientific Acros
190020050
5 g Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10591441
missing translation for 'viewDocuments'
Thermo Scientific Acros
190021000
100 g Glass bottle
32.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
10407760
missing translation for 'viewDocuments'
Thermo Scientific Acros
190025000
500 g Glass bottle
94.97 €
500g
Please to purchase this item. Need a web account? Register with us today!
15179213
missing translation for 'viewDocuments'
Thermo Scientific Acros
190020025
2.5 kg Glass bottle
437.18 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones

Identificadores de productos químicos

112-89-0
333.40
WSULSMOGMLRGKU-UHFFFAOYSA-N
8218
CCCCCCCCCCCCCCCCCCBr
C18H37Br
MFCD00000231
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
1-bromooctadecane

Especificaciones

112-89-0
100.0
Yellow
214.0°C to 216.0°C (12.0 mmHg)
Authentic
C18H37Br
1.4620 to 1.4640
0.97
WSULSMOGMLRGKU-UHFFFAOYSA-N
1-bromooctadecane
8218
96%
1-Bromooctadecane, 96%
95.0
>20.0°C
0.9700g/mL
>110°C
96%
CH3(CH2)17Br
MFCD00000231
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
CCCCCCCCCCCCCCCCCCBr
333.40
333.38
Low Melting Solid
Videos
Seguridad y manipulación

Seguridad y manipulación

missing translation for 'einecsNumber' : 204-013-4

missing translation for 'tsca' : TSCA

Nombre del producto
  • 1-Bromooctadecane

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only