Learn More
1-Bromooctane, 98+%, Thermo Scientific Chemicals
40.90 € - 470.34 €
Chemical Identifiers
CAS | 111-83-1 |
---|---|
Molecular Formula | C8H17Br |
Molecular Weight (g/mol) | 193.13 |
MDL Number | MFCD00000276 |
InChI Key | VMKOFRJSULQZRM-UHFFFAOYSA-N |
Synonym | n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo |
PubChem CID | 8140 |
IUPAC Name | 1-bromooctane |
SMILES | CCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417756
|
Thermo Scientific Alfa Aesar
A14722.30 |
250 g |
40.90 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407756
|
Thermo Scientific Alfa Aesar
A14722.0B |
1000 g |
116.58 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427756
|
Thermo Scientific Alfa Aesar
A14722.0I |
5000 g |
470.34 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and bases.
Chemical Identifiers
111-83-1 | |
193.13 | |
VMKOFRJSULQZRM-UHFFFAOYSA-N | |
8140 | |
CCCCCCCCBr |
C8H17Br | |
MFCD00000276 | |
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo | |
1-bromooctane |
Specifications
111-83-1 | |
1.114 | |
78°C (172°F) | |
C8H17Br | |
1.4518 | |
UN3082 | |
14,6764 | |
Immiscible with water. | |
CCCCCCCCBr | |
193.13 | |
193.13 | |
1-Bromooctane |
-55°C | |
200°C to 202°C | |
Characteristic | |
CH3(CH2)7Br | |
MFCD00000276 | |
1733136 | |
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo | |
VMKOFRJSULQZRM-UHFFFAOYSA-N | |
1-bromooctane | |
8140 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 203-912-9
missing translation for 'rtecsNumber' : RG8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only