Learn More
1-Bromopentadecane, 97%, Thermo Scientific Chemicals
49.55 € - 185.93 €
Chemical Identifiers
CAS | 629-72-1 |
---|---|
Molecular Formula | C15H31Br |
Molecular Weight (g/mol) | 291.32 |
MDL Number | MFCD00000229 |
InChI Key | JKOTZBXSNOGCIF-UHFFFAOYSA-N |
Synonym | pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 |
PubChem CID | 12394 |
IUPAC Name | 1-bromopentadecane |
SMILES | CCCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351987
|
Thermo Scientific Alfa Aesar
L07618.09 |
10 g |
49.55 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361987
|
Thermo Scientific Alfa Aesar
L07618.18 |
50 g |
185.93 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
Chemical Identifiers
629-72-1 | |
291.32 | |
JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
12394 | |
CCCCCCCCCCCCCCCBr |
C15H31Br | |
MFCD00000229 | |
pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 | |
1-bromopentadecane |
Specifications
629-72-1 | |
1.005 | |
>110°C (230°F) | |
1.4608 | |
1744553 | |
Not miscible in water. | |
CCCCCCCCCCCCCCCBr | |
291.32 | |
291.32 | |
1-Bromopentadecane |
18°C to 19°C | |
159°C to 160°C (5 mmHg) | |
C15H31Br | |
MFCD00000229 | |
pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339 | |
JKOTZBXSNOGCIF-UHFFFAOYSA-N | |
1-bromopentadecane | |
12394 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 211-104-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.