missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluorooctane, 98+%, Thermo Scientific Chemicals

Product Code. 11410647
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with oxidizing agents, acids, metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-55-2
Molecular Formula C8BrF17
Molecular Weight (g/mol) 498.97
MDL Number MFCD00042082
InChI Key WTWWXOGTJWMJHI-UHFFFAOYSA-N
Synonym perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide
PubChem CID 9873
ChEBI CHEBI:38803
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Melting Point 6°C
Density 1.93
Boiling Point 141°C to 143°C
Flash Point None
Linear Formula CF3(CF2)7Br
Refractive Index 1.305
Quantity 1 g
Beilstein 2016022
Merck Index 14,7156
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 498.98
Percent Purity ≥98%
Chemical Name or Material 1-Bromoperfluorooctane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'