missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopropane, 99%

Product Code. 10305980 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10305980

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-94-5 | C3H7Br | 122.993 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-94-5
Molecular Formula C3H7Br
MDL Number MFCD00000254
InChI Key CYNYIHKIEHGYOZ-UHFFFAOYSA-N
Synonym n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
PubChem CID 7840
ChEBI CHEBI:47105
IUPAC Name 1-bromopropane
SMILES CCCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2CH2Br
Refractive Index 1.4320 to 1.4360 (20°C, 589nm)
Quantity 250 mL
Molecular Weight (g/mol) 122.993
Formula Weight 122.99
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1-Bromopropane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromopropane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H420-Harms public health and the environment by destroying ozone in the upper atmosphere.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P502-Refer to manufacturer/supplier for information on recovery/recycling.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.