missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopropane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5000 mL
Description
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
Solubility
Miscible with ethanol, water and diethyl ether.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.
Solubility
Miscible with ethanol, water and diethyl ether.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Melting Point | -110°C |
Density | 1.353 |
Boiling Point | 70°C to 71°C |
Flash Point | 21°C (70°F) |
Linear Formula | CH3CH2CH2Br |
Refractive Index | 1.434 |
Quantity | 5000 mL |
UN Number | UN2344 |
Beilstein | 505936 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'