missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Product Code. 13488418
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.28
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 175.0°C to 178.0°C (20.0 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13Br
Refractive Index 1.4590 to 1.4610
Quantity 2.5 kg
Beilstein 01,II,136
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 277.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromotetradecane, 98%
Show More Show Less
Product Identifier
  • 1-Bromotetradecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'