missing translation for 'onlineSavingsMsg'
Learn More
1-Bromotetradecane, 98%, Thermo Scientific Chemicals Product Code.: 11493087

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Product Code. 11493087
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11493087

Brand: Thermo Scientific Alfa Aesar A16082.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.29
MDL Number MFCD00000228
InChI Key KOFZTCSTGIWCQG-UHFFFAOYSA-N
Synonym myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo
PubChem CID 8208
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

Melting Point 4°C to 6°C
Density 1.018
Boiling Point 175°C to 178°C (20 mmHg)
Flash Point 158°C (316°F)
Refractive Index 1.46
Quantity 500 g
Beilstein 1742640
Solubility Information Not miscible in water.
Formula Weight 277.3
Percent Purity 98%
Chemical Name or Material 1-Bromotetradecane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250535 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog NumberA16082.0E,A16082.36,A16082.22
Lot Number10248091 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog NumberA16082.0E,A16082.36,A16082.22
Lot NumberB5158C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16082.0E,A16082.36,A16082.22
Lot Number10227761 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16082.0E,A16082.36,A16082.22
Lot Number10158050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16082.0E,A16082.36,A16082.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromotetradecane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.