missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoundecane, 98%, Thermo Scientific Chemicals

Product Code. 10582811
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-67-4
MDL Number MFCD00000223
InChI Key IKPSIIAXIDAQLG-UHFFFAOYSA-N
Synonym undecyl bromide, undecane, 1-bromo, hendecyl bromide, n-undecyl bromide, bromoundecane, 1-bromo-undecane, n-undecyl-1-bromide, labotest-bb ltbb001165, undecylbromide, 1-bromanylundecane
PubChem CID 12744
IUPAC Name 1-bromoundecane
SMILES CCCCCCCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -9°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 137°C to 138°C (18.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C11H23Br
Linear Formula CH3(CH2)10Br
Refractive Index 1.4555 to 1.4575
Quantity 25 g
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 235.21
Formula Weight 235.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromoundecane, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'