missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10684752 Shop All Thermo Scientific Products
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Butanol, 99%, C4H10O, CAS Number-71-36-3, 1-butanol, 1-hydroxybutane, butanol, butyl alcohol, butyl hydroxide, methylolpropane, n-butanol, n-butyl alcohol, propylcarbinol, propylmethanol, 25L, Vapor Pressure: 6.7mbar at 20 deg.C, 01, 367, 117.6 deg.C, 100.0, 98.5, CHEBI:28885, Undesignated

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropaneShow MoreShow Less
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89.0°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.8100g/mL
Boiling Point 117.6°C
Quantity 1 L
Flash Point 35°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3980 to 1.4000
Assay Percent Range 99%
Residue after Evaporation 0.005% max.
Specific Gravity 0.81
Linear Formula CH3(CH2)3OH
Vapor Pressure Vapor Pressure: 6.7mbar at 20°C
Viscosity 2.95 mPa.s (20°C)
Beilstein 01, 367
Water 0.2% max. (Coulometric)
Merck Index 15, 1542
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
Formula Weight 74.12
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material 1-Butanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468384 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number107690025,107690010,107690250
Lot NumberA0464402 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number107690025,107690010,107690250
Lot NumberA0457560 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number107690025,107690010,107690250
Lot NumberA0396987 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number107690025,107690010,107690250
Lot NumberA0448566 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number107690025,107690010,107690250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.