missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99%, extra pure, Thermo Scientific Chemicals

1-Butanol, 99%, C4H10O, CAS Number-71-36-3, 1-butanol, 1-hydroxybutane, butanol, butyl alcohol, butyl hydroxide, methylolpropane, n-butanol, n-butyl alcohol, propylcarbinol, propylmethanol, 25L, Vapor Pressure: 6.7mbar at 20 deg.C, 01, 367, 117.6 deg.C, 100.0, 98.5, CHEBI:28885, Undesignated

44.10 € - 434.16 €

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10684752
SDS View Documents
Thermo Scientific Acros
107690010
1 L Glass Bottle
44.10 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10376070
SDS View Documents
Thermo Scientific
107690025
2.5 L Glass Bottle
76.82 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10639283
SDS View Documents
Thermo Scientific Acros
107690250
25 L Metal Drum
434.16 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71-36-3
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
263
butan-1-ol
C4H10O
MFCD00002964
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
CHEBI:28885
CCCCO

Specifications

-89.0°C
98.5
0.8100g/mL
35°C
Liquid
99%
0.81
CH3(CH2)3OH
Vapor Pressure: 6.7mbar at 20°C
01, 367
15, 1542
Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
CCCCO
74.123
CHEBI:28885
99%
1-Butanol
71-36-3
100.0
117.6°C
Authentic
1.3980 to 1.4000
0.005% max.
C4H10O
MFCD00002964
2.95 mPa.s (20°C)
0.2% max. (Coulometric)
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
263
74.12
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • 1-Butanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.