missing translation for 'onlineSavingsMsg'
Learn More
1-Butanol, 99%, Thermo Scientific Chemicals Product Code.: 11377946

1-Butanol, 99%, Thermo Scientific Chemicals

Product Code. 11377946
1000 mL
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11377946

Brand: Thermo Scientific Alfa Aesar L13171.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

Solubility
Miscible with alcohols, ketones, aldehydes, ethers, aliphatic and aromatic hydrocarbons. Slightly miscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents, alkali metals and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

pH 7
Melting Point -90°C to -89°C
Density 0.81
Boiling Point 117°C to 118°C
Quantity 1000 mL
Flash Point 35°C (95°F)
Odor Alcohol-like
Refractive Index 1.399
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Percent Purity 99%
Chemical Name or Material 1-Butanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251918 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberL13171.0F,L13171.AE,L13171.AU
Lot Number10225036 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberL13171.0F,L13171.AE,L13171.AU
Lot Number10247589 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberL13171.0F,L13171.AE,L13171.AU
Lot Number10238559 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL13171.0F,L13171.AE,L13171.AU
Lot Number10239956 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL13171.0F,L13171.AE,L13171.AU
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Butanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.