Learn More
1-Butanol, HPLC Grade, 99%, Thermo Scientific Chemicals
Filtered through 0.2μm filters
72.36 € - 242.21 €
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00002964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
Synonym | 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane |
PubChem CID | 263 |
ChEBI | CHEBI:28885 |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378197
|
Thermo Scientific Alfa Aesar
022925.K2 |
1 L |
72.36 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368197
|
Thermo Scientific Alfa Aesar
022925.M1 |
4 x 1 L |
242.21 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Butanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Specifications
−90°C to -89°C | |
0.810 | |
35°C (95°F) | |
1.399 | |
C4H10O | |
969148 | |
Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water. | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
HPLC |
71-36-3 | |
117°C to 118°C | |
Liquid | |
99% | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12 | |
1-Butanol |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H318-H335-H336
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 200-751-6
missing translation for 'rtecsNumber' : EO1400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Butanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1-Butanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.