missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Not available
Poly Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Color | Colorless |
Density | 0.81g/mL at 25°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 0.112 |
Flash Point | 35°C (95°F) |
Ignition Point | 649°F |
Packaging | Not available |
Refractive Index | n20/D 1.399 |
Linear Formula | CH3(CH2)3OH |
UN Number | UN1120 |
Show More |
Safety and Handling
Product Identifier
- 1-Butanol
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction