Learn More
1-Butylamine, 99%, Thermo Scientific Chemicals
23.62 € - 68.34 €
Chemical Identifiers
CAS | 109-73-9 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.139 |
MDL Number | MFCD00011690 |
InChI Key | HQABUPZFAYXKJW-UHFFFAOYSA-N |
Synonym | butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan |
PubChem CID | 8007 |
ChEBI | CHEBI:43799 |
IUPAC Name | butan-1-amine |
SMILES | CCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382216
|
Thermo Scientific Alfa Aesar
L03575.AE |
100 mL |
23.62 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392216
|
Thermo Scientific Alfa Aesar
L03575.AP |
500 mL |
27.14 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302226
|
Thermo Scientific Alfa Aesar
L03575.0F |
2500 mL |
68.34 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N'-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N′-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.
Solubility
Miscible with ethanol and water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, aluminum, copper, copper alloys and acids.
Chemical Identifiers
109-73-9 | |
73.139 | |
HQABUPZFAYXKJW-UHFFFAOYSA-N | |
8007 | |
butan-1-amine |
C4H11N | |
MFCD00011690 | |
butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan | |
CHEBI:43799 | |
CCCCN |
Specifications
109-73-9 | |
0.738 | |
−14°C (6°F) | |
C4H11N | |
1.401 | |
UN1125 | |
Air Sensitive | |
butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan | |
HQABUPZFAYXKJW-UHFFFAOYSA-N | |
butan-1-amine | |
8007 | |
73.14 | |
1-Butylamine |
-50°C | |
76°C to 78°C | |
Ammonia-like | |
CH3CH2CH2CH2NH2 | |
MFCD00011690 | |
605269 | |
14,1543 | |
Miscible with ethanol and water. | |
CCCCN | |
73.139 | |
CHEBI:43799 | |
99% |
Safety and Handling
GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-BUTYLAMINE
missing translation for 'einecsNumber' : 203-699-2
missing translation for 'rtecsNumber' : EO2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Butylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : n-Butylamine
RUO – Research Use Only