Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico 1 & Pico 8 Digital Dispensers
GREENER_CHOICE
The Dispensehead cartidges allow dispensing of one to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution
Brand:  Thermo Scientific™ LTR0001
 View more versions of this product
Product Code. 17961961
  • 1565.00 € / Pack of 30
Explore more special offers
Description
Description
Great choice for dispensing picoliter to microliter volumes for one to eight different types of fluid across an entire plate or just part of a plate.
  • Cost Saving through assay miniaturization
  • Improved efficiency and sample integrity
  • Powerful, flexible, and easy to use software
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital Dispenser Cartridge
1-ch 20μL Cartridges for Multidrop Pico 1
1
Drug combination studies, Biochemical assays, Cell-based assays, DMPK, Secondary screening, Synthetic chemistry, assay development, qPCR, Enzyme profiling, Antibody therapies, SAR, and Spotting
USB
≤ 8%
11 pL to 10 μL in seconds
4-ch 200 μL Cartridges for Pico 8
0.225
100/240 V
Not Autoclavable
1-ch 200μL Cartridges for Pico 1
11 pL to 20 μL, 1 nL to 200 μL
Mulitdrop Pico 1 Digital Dispenser
12, 24, 48, 96, 384, 1536 well plate, Plate height: 6-47 mm
PicoIT 1
8.2 x 6.5 X 2.2 cm
8-ch 20 μL Cartridges for Pico 8
100 to 240V 50/60Hz
50/60 Hz