missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3,4-dinitrobenzene, 90%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
250 g
Packaging:
Glass Bottle
Unit Size:
100g
250g
50g
Specifications
Specifications
| CAS Min % | 88.0 |
| CAS Max % | 100.0 |
| Melting Point | 36.0°C |
| Color | Brown-Yellow |
| Density | 1.6800g/mL |
| Boiling Point | 315.0°C |
| Flash Point | 194°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Packaging | Glass Bottle |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1-Chloro-3,4-dinitrobenzene
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur