missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,5-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Product Code. 11424180
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7051-16-3
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00008382
InChI Key WQHNWJBSROXROL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethoxybenzene, 3,5-dimethoxychlorobenzene, benzene, 1-chloro-3,5-dimethoxy, 5-chlororesorcinol dimethyl ether, 1,3-dimethoxy-5-chlorobenzene, pubchem3042, benzene,5-dimethoxy, acmc-209oev, wln: gr co1 eo1, 3,5-dimethoxyphenyl chloride
PubChem CID 81502
IUPAC Name 1-chloro-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(=C1)Cl)OC

Specifications

Melting Point 32°C to 36°C
Boiling Point 234°C
Quantity 100 g
Beilstein 2327304
Formula Weight 172.61
Percent Purity 98%
Chemical Name or Material 1-Chloro-3,5-dimethoxybenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'