missing translation for 'onlineSavingsMsg'
Learn More

1-chloro-4-(3-iodopropoxy)benzene, 97%, Thermo Scientific™

Product Code. 10099321
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 119795-57-2
Molecular Formula C9H10ClIO
Molecular Weight (g/mol) 296.532
MDL Number MFCD01764552
InChI Key HVVWUBWHLIPSCS-UHFFFAOYSA-N
Synonym 1-chloro-4-3-iodopropoxy benzene, 3-4-chlorophenoxy propyl iodide, benzene,1-chloro-4-3-iodopropoxy, acmc-1c0gn, 1-4-chlorophenoxy-3-iodopropane
PubChem CID 2794890
IUPAC Name 1-chloro-4-(3-iodopropoxy)benzene
SMILES C1=CC(=CC=C1OCCCI)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula ClC6H4OCH2CH2CH2I
Quantity 1 g
Formula Weight 296.53
Percent Purity 97%
Chemical Name or Material 1-chloro-4-(3-iodopropoxy)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'